Learn More
CIAPIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12638-1-AP-150UL
 View more versions of this product
Product Code. 16800464
  • 410.00 € / 150µL
Explore more special offers
Description
Description
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.
Specifications
Specifications
CIAPIN1
Polyclonal
Unconjugated
CIAPIN1
2810413N20Rik, Anamorsin, CIAPIN1, DRE2, PRO0915
Rabbit
Antigen Affinity Chromatography
RUO
109006, 307649, 57019
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XID1, Q6FI81, Q8WTY4
CIAPIN1
CIAPIN1 Fusion Protein Ag3333
150 μL
Primary
Human, Mouse, Rat