missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

33.37 € - 394.97 €

Chemical Identifiers

CAS 17422-74-1
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.155
MDL Number MFCD00014667
InChI Key FSMYWBQIMDSGQP-UHFFFAOYSA-N
Synonym chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde
PubChem CID 87112
IUPAC Name 4-oxochromene-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11432787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15438.06
5 g
33.37 €
5g
Please to purchase this item. Need a web account? Register with us today!
11442787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15438.14
25 g
106.53 €
25g
Please to purchase this item. Need a web account? Register with us today!
11452787
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15438.22
100 g
394.97 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17422-74-1
174.155
FSMYWBQIMDSGQP-UHFFFAOYSA-N
87112
C1=CC=C2C(=C1)C(=O)C(=CO2)C=O
C10H6O3
MFCD00014667
chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde
4-oxochromene-3-carbaldehyde

Specifications

17422-74-1
C10H6O3
472849
chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde
C1=CC=C2C(=C1)C(=O)C(=CO2)C=O
174.155
174.16
Chromone-3-carboxaldehyde
150°C to 154°C
MFCD00014667
Air sensitive
FSMYWBQIMDSGQP-UHFFFAOYSA-N
4-oxochromene-3-carbaldehyde
87112
97%
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 241-451-5

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only