missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(Vi) Oxide, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Specifications
Melting Point | 196°C |
Color | Red to Violet |
pH | 1 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 99.99g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Chromium(VI) oxide
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Oxidising solid Category 1
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 A
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H350-May cause cancer.
- H340-May cause genetic defects.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H361f-Suspected of damaging fertility.
- H301-Toxic if swallowed.
- H271-May cause fire or explosion; strong oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction