missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
96.40 € - 213.00 €
Chemical Identifiers
CAS | 7789-09-5, 7732-18-5, 7789-09-5 |
---|---|
Molecular Formula | Cr2H8N2O7 |
Molecular Weight (g/mol) | 252.06 |
InChI Key | JOSWYUNQBRPBDN-UHFFFAOYSA-P |
PubChem CID | 24600 |
IUPAC Name | diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10086563
|
SPEX Certiprep
PLCR2-3Y |
125 mL | Triple-leached LDPE bottle |
96.40 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10000482
|
SPEX Certiprep
PLCR2-3X |
500 mL | Triple-leached LDPE bottle |
213.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cr | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7789-09-5, 7732-18-5, 7789-09-5 | |
252.06 | |
24600 | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Cr2H8N2O7 | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
7789-09-5, 7732-18-5, 7789-09-5 | |
0.1, 2.0, 97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Cr2H8N2O7 | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
252.06 | |
Assurance | |
5% HNO3 |
0.1, 2.0, 97.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24600 | |
Matrix Concentration Cat. No.2-5% HNO3 1000μg/mL PLCR2-2Y, -2T, -2X 10,000 PLCR2-3Y, -3X2-5% HCl 1000μg/mL PLCR1-2X 10,000 PLCR1-3XH2O 1000μg/mL PLCR9-2X 10,000 PLCR9-3X | |
Chromium Standard |
Safety and Handling
May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. Causes serious eye irritation
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 10,000
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : chromium(III) nitrate nonahydrate, nitric acid
Product intended for laboratory use only