Learn More
Chromium powder, -100 mesh, 99% (metals basis), Thermo Scientific Chemicals
Apparent density 2.0-3.0g/cm3
34.77 € - 1160.78 €
Chemical Identifiers
CAS | 7440-47-3 |
---|---|
Molecular Formula | Cr |
Molecular Weight (g/mol) | 52.00 |
MDL Number | MFCD00010944 |
InChI Key | VYZAMTAEIAYCRO-UHFFFAOYSA-N |
Synonym | chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 |
PubChem CID | 23976 |
ChEBI | CHEBI:28073 |
IUPAC Name | chromium |
SMILES | [Cr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361198
|
Thermo Scientific Alfa Aesar
010147.09 |
10 g |
34.77 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381198
|
Thermo Scientific Alfa Aesar
010147.22 |
100 g |
98.19 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371198
|
Thermo Scientific Alfa Aesar
010147.36 |
500 g |
360.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391198
|
Thermo Scientific Alfa Aesar
010147.A3 |
2 kg |
1160.78 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-47-3 | |
52.00 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
23976 | |
chromium |
Cr | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
CHEBI:28073 | |
[Cr] |
Specifications
Gray | |
Gray powder | |
Cr | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
[Cr] | |
52.00 | |
CHEBI:28073 | |
Odorless | |
Chromium |
7440-47-3 | |
99% (Metals basis) | |
MFCD00010944 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
chromium | |
23976 | |
-100 Mesh | |
Powder |
Safety and Handling
Wear suitable protective clothing and gloves.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-157-5
missing translation for 'rtecsNumber' : GB4200000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Chromium powder
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
- Carcinogenicity Category 2
Hazard Statement
- H228-Flammable solid.
- H351-Suspected of causing cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only