missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Oxide, Green, Extra Pure, SLR, Fisher Chemical

Product Code. 10714371
Click to view available options
500g, Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Boiling Point 4000°C
Melting Point 2435°C
CAS 1308-38-9
Color Green
pH 5 to 7
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula Cr2O3
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]
IUPAC Name dichromium(3+) trioxidandiide
Molecular Weight (g/mol) 151.99
Formula Weight 151.99g/mol
Packaging Plastic powder jar
Show More Show Less
Product Identifier
  • Chromium(III) oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'