Learn More
Chromium(III) fluoride hydrate, Thermo Scientific™
44.00 € - 129.65 €
Chemical Identifiers
CAS | 123333-98-2 |
---|---|
Molecular Formula | CrF3 |
Molecular Weight (g/mol) | 108.99 |
MDL Number | MFCD00016045 |
InChI Key | FTBATIJJKIIOTP-UHFFFAOYSA-K |
IUPAC Name | chromium(3+) trifluoride |
SMILES | [F-].[F-].[F-].[Cr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351709
|
Thermo Scientific Alfa Aesar
012339.30 |
250 g | N/A | ||||||
11341709
|
Thermo Scientific Alfa Aesar
012339.A1 |
1 kg |
129.65 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
123333-98-2 | |
108.99 | |
FTBATIJJKIIOTP-UHFFFAOYSA-K | |
[F-].[F-].[F-].[Cr+3] |
CrF3 | |
MFCD00016045 | |
chromium(3+) trifluoride |
Specifications
>1000°C | |
Green | |
CrF3 | |
UN1756 | |
FTBATIJJKIIOTP-UHFFFAOYSA-K | |
chromium(3+) trifluoride | |
108.99 (Anhydrous) | |
Chromium(III) fluoride hydrate |
123333-98-2 | |
Crystalline | |
MFCD00016045 | |
14,2223 | |
[F-].[F-].[F-].[Cr+3] | |
108.99 | |
Hygroscopic |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHROMIC FLUORIDE, SOLID
missing translation for 'tsca' : TSCA