missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) fluoride, 98%, Thermo Scientific Chemicals

Product Code. 11348617
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-97-8
MDL Number MFCD00016045
Synonym chromium fluoride, chromic fluoride, chromium 3+ trifluoride, acmc-1bht2, chromium 3+ ion trifluoride, chromic fluoride, solid un1756 corrosive, chromic fluoride, solution un1757 corrosive

Specifications

Melting Point >1000°C
Color Green
Physical Form Powder
Quantity 10 g
UN Number UN1756
Merck Index 14,2223
Formula Weight 108.99
Percent Purity 99.98%
Odor Odorless
Sensitivity Hygroscopic
Density 3.8 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(III) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'