missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

 

62.91 € - 159.80 €

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11310939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010686.06
5 g
62.91 €
5g
Please to purchase this item. Need a web account? Register with us today!
11300939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010686.14
25 g
159.80 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chromium(III) chloride hexahydrate is used as an organic synthesis catalyst and as an additive to intravenous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

10060-12-5
266.44
LJAOOBNHPFKCDR-UHFFFAOYSA-K
104957
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Cl3CrH12O6
MFCD00149660
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
CHEBI:53442

Specifications

83°C
Green
Cl3CrH12O6
MFCD00149660
14,2222
Soluble in water and alcohol. Insoluble in diethyl ether.
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
266.44
CHEBI:53442
99.998%
Odorless
(metals basis)
10060-12-5
Crystalline Aggregates
CrCl3·6H2O
UN3260
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium(3+) hexahydrate trichloride
104957
266.45 (158.35 Anhydrous)
Puratronic™
1.835 g/mL
Chromium(III) chloride hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c

H290-H302-H317

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 233-038-3

missing translation for 'rtecsNumber' : GB5450000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only