missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) acetate hydroxide, Cr 24%, Thermo Scientific Chemicals

44.32 € - 108.54 €

Chemical Identifiers

CAS 39430-51-8
Molecular Formula Cr3(OH)2(OOCCH3)7
MDL Number MFCD00058912
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331849
View Documents
Thermo Scientific Alfa Aesar
012518.36
500 g
44.32 €
500g
Please to purchase this item. Need a web account? Register with us today!
11321849
View Documents
Thermo Scientific Alfa Aesar
012518.A3
2 kg
108.54 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39430-51-8
Cr3(OH)2(OOCCH3)7
MFCD00058912

Specifications

39430-51-8
Powder
MFCD00058912
603.31
Chromium(III) acetate hydroxide, Cr 24%
Blue
Cr3(OH)2(OOCCH3)7
Soluble in water. Insoluble in acetone. Soluble in hot water
Odorless
Videos
Safety and Handling

Safety and Handling

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'einecsNumber' : 254-447-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.