missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.
Solubility
Soluble in water. Insoluble in alcohol and ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, water and active metals.
Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.
Solubility
Soluble in water. Insoluble in alcohol and ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, water and active metals.
Specifications
Specifications
Melting Point | 824°C |
Quantity | 25 g |
UN Number | UN3260 |
Merck Index | 14,2244 |
Solubility Information | Soluble in water. Insoluble in alcohol and ether. |
Formula Weight | 122.9 |
Percent Purity | 97% |
Odor | Odorless |
Sensitivity | Air and moisture sensitive |
Density | 2.878 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'