Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 8
Highly pure paper for blotting, chromatography, and a wide range of life science and diagnostics absorption applications.
Brand:  Sartorius Stedim Biotech™ FT-2-509-470580K
Product Code. 15942800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Highly pure paper grade.
  • Ideal for blotting and chromatography.
  • Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Non-sterile, bulk packed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk, 280 g/m2
0.9 mm
FN 8
470 mm × 580 mm
Sheet
Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash)