Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 6
Highly pure paper for blotting, chromatography, and a wide range of life science and diagnostics absorption applications.
Brand:  Sartorius Stedim Biotech™ FT-2-506-210297N
Product Code. 15922800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Highly pure paper grade.
  • Ideal for blotting and chromatography.
  • Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Non-sterile, bulk packed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100/Pk, 125 g/m2
0.22 mm
FN 6
210 mm × 297 mm
Sheet
Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash)