missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carbonyl chloride, 95%, Thermo Scientific™

113.00 € - 275.00 €

Chemical Identifiers

CAS 55746-03-7
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD12198108
InChI Key DKSNWGZAMLPCLZ-UHFFFAOYSA-N
Synonym chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro
PubChem CID 10442680
IUPAC Name 3,4-dihydro-2H-chromene-6-carbonyl chloride
SMILES C1CC2=C(C=CC(=C2)C(=O)Cl)OC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10113034
View Documents
Thermo Scientific Maybridge
CC32802CB
250 mg Amber Glass Bottle
113.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10173074
View Documents
Thermo Scientific Maybridge
CC32802DA
1 g Amber Glass Bottle
275.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

55746-03-7
196.63
DKSNWGZAMLPCLZ-UHFFFAOYSA-N
10442680
C1CC2=C(C=CC(=C2)C(=O)Cl)OC1
C10H9ClO2
MFCD12198108
chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro
3,4-dihydro-2H-chromene-6-carbonyl chloride

Specifications

55746-03-7
100.0
C10H9ClO2
chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro
C1CC2=C(C=CC(=C2)C(=O)Cl)OC1
196.63
196.63
Chroman-6-carbonyl chloride
94.5
95%
MFCD12198108
DKSNWGZAMLPCLZ-UHFFFAOYSA-N
3,4-dihydro-2H-chromene-6-carbonyl chloride
10442680
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.