Learn More
Macherey-Nagel™ Chromabond Florisil Adsorbent
Brand:  Macherey-Nagel™ 730622
Product Code. 12887491
  • 129.00 € / 100g
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
100 g
Organic Tin Compounds, Aliphatic Carboxylic Acids, PCBs, PAHs
Chromabond Florisil Adsorbent
150 to 250 μm