missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Bromobutyl Rubber Injection Stopper in Ready to Sterilize Bag

Provides excellent stability during the freeze-drying process. Brombutyl Rubber Injection Stoppers in Ready to Sterilize Bag are compatible with neutral glass vials.

Brand:  Azpack™ 10744955

Product Code. 10744955

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 20mm freeze drying stoppers in ready-to-sterilise bag
  • For use wtih 5mL, 10mL and 20mL vials
Specifications

Specifications

Stopper
2000/Pk.
20 mm
Bromobutyl Rubber
Compatible with VGA-220-121U, -132P and -241K 5mL, 10mL and 20mL vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.