missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | CHRNA10 |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836785
|
Proteintech
55291-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16826785
|
Proteintech
55291-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to CHRNA10.
Ionotropic receptor with a probable role in the modulation of auditory stimuli. Agonist binding may induce an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane. The channel is permeable to a range of divalent cations including calcium, the influx of which may activate a potassium current which hyperpolarizes the cell membrane. In the ear, this may lead to a reduction in basilar membrane motion, altering the activity of auditory nerve fibers and reducing the range of dynamic hearing. This may protect against acoustic trauma.Specifications
CHRNA10 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q9GZZ6, Q9JLB5 | |
504186, 57053, 64574 | |
Peptide | |
Primary | |
-20°C | |
CHRNA10 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CHRNA10, NACHR alpha 10, NACHRA10 | |
Chrna10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |