missing translation for 'onlineSavingsMsg'
Learn More

Choline iodide, 98%, Thermo Scientific Chemicals

Product Code. 11324334
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17773-10-3
Molecular Formula C5H14INO
Molecular Weight (g/mol) 231.08
MDL Number MFCD00011900
InChI Key FNPBHXSBDADRBT-UHFFFAOYSA-M
Synonym choline iodide, jodoetano, dilatol zambon, 2-hydroxy-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, iodide, 2-hydroxyethyl trimethylammonium iodide, cholineiodide, acmc-209ed9, d0q2de, 2-hydroxyethyl trimethyl azanium iodide
PubChem CID 87300
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;iodide
SMILES [I-].C[N+](C)(C)CCO

Specifications

Melting Point ∼270°C
Quantity 50 g
Beilstein 3912691
Formula Weight 231.08
Percent Purity 98%
Chemical Name or Material Choline iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'