missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotri-n-butylsilane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications
Specifications
Boiling Point | 93°C to 94°C (4mmHg) |
Quantity | 25 g |
UN Number | UN2987 |
Beilstein | 1742537 |
Flash Point | 104°C (219°F) |
Solubility Information | Reacts with water. |
Formula Weight | 234.89 |
Percent Purity | 97% |
Refractive Index | 1.446 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction