missing translation for 'onlineSavingsMsg'
Learn More

Chlorotri-n-butylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11312176
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotri-n-butylsilane is as silylating agent. It can react with bromo-trifluoro-methane, to product tributyl-trifluoromethyl-silane.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 995-45-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00000509
InChI Key JSQJUDVTRRCSRU-UHFFFAOYSA-N
Synonym silane, tributylchloro, chlorotributylsilane, chlorotri-n-butylsilane, tributyl chloro silane, tri-n-butylchlorosilane, tributylsilyl chloride, tributylchloro-silan, chloro-tri-n-butylsilane
PubChem CID 70448
IUPAC Name tributyl(chloro)silane
SMILES CCCC[Si](CCCC)(CCCC)Cl

Specifications

Boiling Point 93°C to 94°C (4mmHg)
Quantity 5 g
UN Number UN2987
Beilstein 1742537
Flash Point 104°C (219°F)
Solubility Information Reacts with water.
Formula Weight 234.89
Percent Purity 97%
Refractive Index 1.446
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material Chlorotri-n-butylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'