missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine Diphosphate Salt, MP Biomedicals

Product Code. 11426331
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18H32ClN3O8P2
Molecular Weight (g/mol) 515.86
MDL Number MFCD00069852
InChI Key QKICWELGRMTQCR-UHFFFAOYNA-N
Synonym chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
PubChem CID 64927
SMILES OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

Specifications

Quantity 25 g
IUPAC Name 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid
Formula Weight 515.9
Chemical Name or Material Chloroquine Diphosphate Salt
compliance-icons
Product Identifier
  • Chloroquine Diphosphate Salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Chloroquinone Phosphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'