missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Honeywell™
ACS Reagent,≥37.50% Pt basis
286.59 € - 2622.00 €
Chemical Identifiers
CAS | 18497-13-7 |
---|---|
Molecular Formula | Cl6H14O6Pt |
Molecular Weight (g/mol) | 517.89 |
MDL Number | MFCD00149910 |
InChI Key | PIJUVEPNGATXOD-UHFFFAOYSA-J |
IUPAC Name | platinum(4+) dihydrogen hexahydrate hexachloride |
SMILES | [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
15671770
|
Honeywell-Fluka
206083-1G |
1 g | Glass Bottle |
286.59 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15691770
|
Honeywell-Fluka
206083-5G |
5 g | Glass Bottle | N/A | N/A | N/A | |||||
15681770
|
Honeywell-Fluka
206083-25G |
25 g | Glass Bottle | N/A | N/A | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
18497-13-7 | |
517.89 | |
PIJUVEPNGATXOD-UHFFFAOYSA-J | |
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4] |
Cl6H14O6Pt | |
MFCD00149910 | |
platinum(4+) dihydrogen hexahydrate hexachloride |
Specifications
18497-13-7 | |
Cl6H14O6Pt | |
MFCD00149910 | |
PIJUVEPNGATXOD-UHFFFAOYSA-J | |
platinum(4+) dihydrogen hexahydrate hexachloride | |
517.9g/mol | |
ACS Reagent |
Powder | |
H2PtCl6 · 6H2O | |
UN2507 | |
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4] | |
517.89 | |
≥37.50% (Pt basis) | |
Chloroplatinic Acid Hexahydrate |
Safety and Handling
P261-P280-P301 + P310-P305 + P351 + P338-P310
H301-H314-H317-H334
missing translation for 'dotInformation' : 8/PGIII
missing translation for 'einecsNumber' : 241-010-7
missing translation for 'rtecsNumber' : TP1510000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Chloroplatinic acid hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Chloroplatinic acidhexahydrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Honeywell™