missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 11442487
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

Solubility
Soluble in water (0.4g/L at 20°C).

Notes
Store in cool, well ventilated area. Keep container tightly closed. Moisture sensitive. Store under argon.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2344-80-1
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00000878
InChI Key OOCUOKHIVGWCTJ-UHFFFAOYSA-N
Synonym chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane
PubChem CID 75361
IUPAC Name chloromethyl(trimethyl)silane
SMILES C[Si](C)(C)CCl

Specifications

Boiling Point 97°C to 98°C
Quantity 50 g
UN Number UN1993
Beilstein 906705
Flash Point −2°C (28°F)
Solubility Information Soluble in water (0.4g/L at 20°C).
Formula Weight 122.67
Percent Purity 98%
Odor Odorless
Refractive Index 1.418
Sensitivity Moisture sensitive
Density 0.886
Chemical Name or Material (Chloromethyl)trimethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'