Learn More
Chloroform-d, 99.8% (Isotopic), Thermo Scientific Chemicals
47.40 € - 160.00 €
Chemical Identifiers
CAS | 865-49-6 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 120.375 |
MDL Number | MFCD00000827 |
InChI Key | HEDRZPFGACZZDS-MICDWDOJSA-N |
Synonym | chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero |
PubChem CID | 71583 |
ChEBI | CHEBI:85365 |
IUPAC Name | trichloro(deuterio)methane |
SMILES | C(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403800
|
Thermo Scientific Alfa Aesar
089541.18 |
50 g |
47.40 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413800
|
Thermo Scientific Alfa Aesar
089541.30 |
250 g |
160.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chloroform-d is widely used in the organic solvent for the NMR analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
865-49-6 | |
120.375 | |
HEDRZPFGACZZDS-MICDWDOJSA-N | |
71583 | |
trichloro(deuterio)methane |
CHCl3 | |
MFCD00000827 | |
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero | |
CHEBI:85365 | |
C(Cl)(Cl)Cl |
Specifications
Liquid | |
1.50 g/mL | |
Light and Moisture | |
-64°C | |
CHCl3 | |
UN1888 | |
Miscible with organic solvents ethyl acetate,acetone. | |
C(Cl)(Cl)Cl | |
120.375 | |
CHEBI:85365 | |
99.8% |
Colorless | |
1.4445 | |
865-49-6 | |
62°C | |
MFCD00000827 | |
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero | |
HEDRZPFGACZZDS-MICDWDOJSA-N | |
trichloro(deuterio)methane | |
71583 | |
120.38 | |
Chloroform-d, Isotopic |
Safety and Handling
GHS H Statement
H351-H373-H302-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P362-P501c
H302-H315-H319-H331-H336-H351-H361d-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROFORM
missing translation for 'einecsNumber' : 212-742-4
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only