missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, CHROMASOLV™, for HPLC, ≥99.8%, amylene stabilized, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC,≥99.8%, amylene stabilized
70.76 € - 873.65 €
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
IUPAC Name | trichloromethane |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15634530
|
Honeywell-Chromasolv
34854-100ML |
100 mL | Glass Bottle | ||||||
15644530
|
Honeywell-Chromasolv
34854-1L |
1 L | Glass Bottle |
118.34 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15664530
|
Honeywell-Chromasolv
34854-2L |
2 L | Glass Bottle | ||||||
15654530
|
Honeywell-Chromasolv
34854-2.5L |
2.5 L | Glass Bottle |
244.34 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15684530
|
Honeywell-Chromasolv
34854-6X1L |
6 x 1 L | Glass Bottle |
600.25 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15674530
|
Honeywell-Chromasolv
34854-4X2.5L |
4 x 2.5 L | Glass Bottle |
873.65 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
trichloromethane |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 | |
ClC(Cl)Cl |
Specifications
-63°C (literature) | |
67-66-3 | |
≤100.0000% | |
CHCl3 | |
UN1888 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
>99.8% | |
1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.) | |
Chloroform, CHROMASOLV™, for HPLC, ≥99.8%, amylene stabilized |
max. 1.00 at 250 nm, max. 0.30 at 260 nm, max. 0.005 280 nm | |
≥90.0000% | |
CHCl3 | |
MFCD00000826 | |
1731042 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38 | |
HPLC | |
61°C |
Safety and Handling
missing translation for 'shelfLife' : 720 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
missing translation for 'einecsNumber' : 200-663-8
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Chloroform
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.