missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorodiphenylsilane, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
Specifications
Specifications
Boiling Point | 280°C |
Quantity | 5 g |
UN Number | UN2987 |
Beilstein | 2937164 |
Flash Point | 105°C (221°F) |
Solubility Information | Reacts with water. |
Formula Weight | 218.76 |
Percent Purity | 90% |
Grade | Technical |
Refractive Index | 1.579 |
Show More |
Safety and Handling
Product Identifier
- Chlorodiphenylsilane
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Chlorodiphenylsilane
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'