missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylsilane, tech. 90%, Thermo Scientific Chemicals

Product Code. 11302236
Click to view available options
Quantity:
5 g
25 g

Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodiphenylsilane is used in Medicine field, electronics industry, polymer materials. It is used in reduction of alcohols.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from bases, water. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1631-83-0
Molecular Formula C12H10ClSi
Molecular Weight (g/mol) 217.747
MDL Number MFCD00045144
InChI Key YCITZMJNBYYMJO-UHFFFAOYSA-N
Synonym chlorodiphenylsilane, diphenylchlorosilane, chloro diphenyl silane, silane, chlorodiphenyl, benzene, 1,1'-chlorosilylene bis, c12h11clsi, chlorodiphenylsilyl, diphenylsilyl chloride, chloro diphenyl silicon
PubChem CID 6327323
IUPAC Name chloro(diphenyl)silicon
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)Cl

Specifications

Boiling Point 280°C
Quantity 25 g
UN Number UN2987
Beilstein 2937164
Flash Point 105°C (221°F)
Solubility Information Reacts with water.
Formula Weight 218.76
Percent Purity 90%
Grade Technical
Refractive Index 1.579
Sensitivity Moisture sensitive
Density 1.123
Chemical Name or Material Chlorodiphenylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'