Learn More
Chlorodimethylsilane, 97%, Thermo Scientific Chemicals
26.60 € - 183.00 €
Chemical Identifiers
CAS | 1066-35-9 |
---|---|
Molecular Formula | C2H6ClSi |
Molecular Weight (g/mol) | 93.61 |
MDL Number | MFCD00000495 |
InChI Key | QABCGOSYZHCPGN-UHFFFAOYSA-N |
Synonym | chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical |
PubChem CID | 6327132 |
IUPAC Name | chloro(dimethyl)silicon |
SMILES | C[Si](C)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15433067
|
Thermo Scientific Alfa Aesar
A13113.AB |
10 mL |
26.60 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443067
|
Thermo Scientific Alfa Aesar
A13113.AD |
50 mL |
56.70 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453067
|
Thermo Scientific Alfa Aesar
A13113.AK |
250 mL |
183.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.
Solubility
Miscible with organic solvents.
Notes
It is incompatible with strong oxidizing agents, strong acids, strong bases, metals and water.
Chemical Identifiers
1066-35-9 | |
93.61 | |
QABCGOSYZHCPGN-UHFFFAOYSA-N | |
6327132 | |
C[Si](C)Cl |
C2H6ClSi | |
MFCD00000495 | |
chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical | |
chloro(dimethyl)silicon |
Specifications
34°C to 36°C | |
1066-35-9 | |
(CH3)2SiHCl | |
UN2924 | |
−28°C (−18°F) | |
QABCGOSYZHCPGN-UHFFFAOYSA-N | |
chloro(dimethyl)silicon | |
6327132 | |
97% | |
Chlorodimethylsilane |
-111°C | |
C2H6ClSi | |
MFCD00000495 | |
1633434 | |
chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical | |
C[Si](C)Cl | |
93.61 | |
94.62 | |
Acrid |
Safety and Handling
GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P280-P305+P351+P338-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'tsca' : TSCA
Recommended Storage : 2° to 8°C
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 A
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H261-In contact with water releases flammable gases.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only