Learn More
Chlorocyclohexane, 98%, Thermo Scientific Chemicals
36.88 € - 121.61 €
Chemical Identifiers
CAS | 542-18-7 |
---|---|
Molecular Formula | C6H11Cl |
Molecular Weight (g/mol) | 118.604 |
MDL Number | MFCD00003822 |
InChI Key | UNFUYWDGSFDHCW-UHFFFAOYSA-N |
Synonym | cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane |
PubChem CID | 10952 |
ChEBI | CHEBI:39156 |
IUPAC Name | chlorocyclohexane |
SMILES | C1CCC(CC1)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356896
|
Thermo Scientific Alfa Aesar
L04349.30 |
250 g |
36.88 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366896
|
Thermo Scientific Alfa Aesar
L04349.0B |
1000 g |
121.61 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.
Solubility
Soluble in water.0.02 g/L (20°C). Soluble in alcohol and diethyl ether.
Notes
Incompatible materials are Strong oxidizing agents, Strong bases.
Chemical Identifiers
542-18-7 | |
118.604 | |
UNFUYWDGSFDHCW-UHFFFAOYSA-N | |
10952 | |
chlorocyclohexane |
C6H11Cl | |
MFCD00003822 | |
cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane | |
CHEBI:39156 | |
C1CCC(CC1)Cl |
Specifications
542-18-7 | |
1 | |
28°C (82°F) | |
C6H11Cl | |
MFCD00003822 | |
1900796 | |
cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane | |
UNFUYWDGSFDHCW-UHFFFAOYSA-N | |
chlorocyclohexane | |
10952 | |
118.61 | |
Chlorocyclohexane |
-44°C | |
141°C to 143°C | |
Unpleasant | |
1.462 | |
UN1993 | |
14,2732 | |
Soluble in water.0.02g/L (20°C). Soluble in alcohol and diethyl ether. | |
C1CCC(CC1)Cl | |
118.604 | |
CHEBI:39156 | |
98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 208-806-6
missing translation for 'rtecsNumber' : GU8685000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only