Learn More
Chlorobis(cyclooctene)rhodium(I) dimer, 98%, Thermo Scientific Chemicals
56.40 € - 228.00 €
Chemical Identifiers
CAS | 12279-09-3 |
---|---|
Molecular Formula | C32H56Cl2Rh2 |
Molecular Weight (g/mol) | 717.51 |
MDL Number | MFCD00013287 |
InChI Key | GQPAPAIPOLEZHT-XFCUKONHSA-L |
Synonym | chlorobis cyclooctene rhodium i dimer, chlorobis cyclooctene rhodium dimer |
PubChem CID | 53384308 |
IUPAC Name | cyclooctene;rhodium;dichloride |
SMILES | [Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10008881
|
Thermo Scientific Acros
369371000 |
100 mg | Glass bottle |
56.40 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10602233
|
Thermo Scientific Acros
369375000 |
500 mg | Glass bottle |
228.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12279-09-3 | |
717.51 | |
GQPAPAIPOLEZHT-XFCUKONHSA-L | |
53384308 | |
[Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1 |
C32H56Cl2Rh2 | |
MFCD00013287 | |
chlorobis cyclooctene rhodium i dimer, chlorobis cyclooctene rhodium dimer | |
cyclooctene;rhodium;dichloride |
Specifications
12279-09-3 | |
98% | |
MFCD00013287 | |
Solubility in water: insoluble. | |
[Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1 | |
717.51 | |
717.52 | |
Chlorobis(cyclooctene)rhodium(I) dimer |
187.0°C to 191.0°C | |
C32H56Cl2Rh2 | |
chlorobis cyclooctene rhodium i dimer, chlorobis cyclooctene rhodium dimer | |
GQPAPAIPOLEZHT-XFCUKONHSA-L | |
cyclooctene;rhodium;dichloride | |
53384308 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- Chlorobis(cyclooctene)rhodium(I) dimer
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only