missing translation for 'onlineSavingsMsg'
Learn More
Chloroauric Acid, Extra Pure, about 51% Au, SLR, Fisher Chemical Product Code.: 10765131

Chloroauric Acid, Extra Pure, about 51% Au, SLR, Fisher Chemical

Product Code. 10765131
1g, Vial
Click to view available options
1g, Vial
This item is not returnable. View return policy

Product Code. 10765131

Brand: Fisher Chemical C/4640/43

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 30°C
CAS 16903-35-8
Color Orange
Physical Form Solid
Quantity 1 g
Molecular Formula HAuCl4
MDL Number 149903
Formula Weight 339.78g/mol
Packaging Vial
compliance-icons compliance-icons
Product Identifier
  • Hydrogen tetrachloroaurate(III) hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroauric Acid, Extra Pure, about 51% Au, SLR, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.