Learn More
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Thermo Scientific Chemicals
214.07 € - 531.65 €
Chemical Identifiers
CAS | 12092-47-6 |
---|---|
Molecular Formula | C16H24Cl2Rh2 |
Molecular Weight (g/mol) | 493.08 |
MDL Number | MFCD00012415 |
InChI Key | QSUDXYGZLAJAQU-MIXQCLKLSA-L |
Synonym | chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer |
PubChem CID | 6436379 |
IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride |
SMILES | [Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375968
|
Thermo Scientific Alfa Aesar
010466.MD |
250 mg |
214.07 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365968
|
Thermo Scientific Alfa Aesar
010466.03 |
1 g |
531.65 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is a widely used precursor to homogeneous catalysts. This is a chiral catalyst capable of asymmetrically hydrogenating certain prochiral alkenes. Chloro(1,5-cyclooctadiene)rhodium(I) dimer is also used in the synthesis of other metal ligands for use in catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12092-47-6 | |
493.08 | |
QSUDXYGZLAJAQU-MIXQCLKLSA-L | |
6436379 | |
[Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
C16H24Cl2Rh2 | |
MFCD00012415 | |
chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer | |
(1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride |
Specifications
12092-47-6 | |
Yellow to Orange | |
C16H24Cl2Rh2 | |
Air sensitive; Hygroscopic | |
Soluble in chloroform,dichloromethane,and methanol; Sparingly soluble in most common solvents. | |
[Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 | |
493.08 | |
493.08 | |
Chloro(1,5-cyclooctadiene)rhodium(I) dimer |
243°C (decomposition) | |
Odorless | |
MFCD00012415 | |
chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer | |
QSUDXYGZLAJAQU-MIXQCLKLSA-L | |
(1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride | |
6436379 | |
Crystalline |
Safety and Handling
missing translation for 'einecsNumber' : 235-157-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only