Learn More
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh, Thermo Scientific Chemicals
522.00 € - 4160.00 €
Chemical Identifiers
CAS | 12092-47-6 |
---|---|
Molecular Formula | C16H24Cl2Rh2 |
Molecular Weight (g/mol) | 493.08 |
MDL Number | MFCD00012415 |
InChI Key | QSUDXYGZLAJAQU-MIXQCLKLSA-L |
Synonym | chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer |
PubChem CID | 6436379 |
IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride |
SMILES | [Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10266600
|
Thermo Scientific Acros
211305000 |
500 mg | Glass bottle |
522.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10492845
|
Thermo Scientific Acros
211300050 |
5 g | Glass bottle |
4160.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12092-47-6 | |
493.08 | |
QSUDXYGZLAJAQU-MIXQCLKLSA-L | |
6436379 | |
[Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
C16H24Cl2Rh2 | |
MFCD00012415 | |
chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer | |
(1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride |
Specifications
12092-47-6 | |
Authentic | |
MFCD00012415 | |
Solubility in water: 0.34g/L | |
[Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 | |
493.08 | |
493.08 | |
Crystalline Powder or Crystals |
256.0°C | |
C16H24Cl2Rh2 | |
chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer | |
QSUDXYGZLAJAQU-MIXQCLKLSA-L | |
(1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride | |
6436379 | |
≥40.8% (RH) | |
Chloro(1, 5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 235-157-6
missing translation for 'tsca' : TSCA
RUO – Research Use Only