missing translation for 'onlineSavingsMsg'
Learn More

chloro(1,5-cyclooctadiene)iridium(I) dimer, Thermo Scientific Chemicals

Product Code. 10704104
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12112-67-3
Molecular Formula C16H24Cl2Ir2
Molecular Weight (g/mol) 671.70
MDL Number MFCD00012414
InChI Key ZFOUDQNHNLDNLD-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene iridium i dimer, bis 1,5-cyclooctadiene diiridium i dichloride, 1z,5z-cycloocta-1,5-diene; iridium; dichloride, iridium, di-.mu.-chlorobis 1,2,5,6-.eta.-1,5-cyclooctadiene di, ir2cl2 cod 2, chloro-1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; iridium 1+
PubChem CID 6436381
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;dichloride
SMILES [Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 190.0°C
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 671.68
Chemical Name or Material chloro(1, 5-cyclooctadiene)iridium(I) dimer
compliance-icons
Product Identifier
  • Chloro(1,5-cyclooctadiene)iridium(I) dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'