missing translation for 'onlineSavingsMsg'
Learn More

Chloral Hydrate, Extra Pure, SLR, Eur.Ph., BP, Fisher Chemical

Product Code. 10690321
Click to view available options
500g, Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 302-17-0
Melting Point 57°C
Color White
pH 3.5 to 5.5
Boiling Point 97°C
Packaging Plastic powder jar
Molecular Formula C2H3Cl3O2
MDL Number 44479
Quantity 500 g
Formula Weight 165.4g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Chloral hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.