missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHI3L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12036-1-AP-20UL
This item is not returnable.
View return policy
Description
Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.Specifications
CHI3L1 | |
Polyclonal | |
Unconjugated | |
CHI3L1 | |
39 kDa synovial protein, ASRT7, Cartilage glycoprotein 39, CGP 39, CHI3L1, Chitinase 3 like protein 1, GP 39, GP39, HC gp39, hCGP 39, HCGP 3P, YKL 40, YKL40, YYL 40 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1116, 12654, 89824 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36222, Q61362, Q9WTV1 | |
Chi3l1 | |
CHI3L1 Fusion Protein Ag2710 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction