Learn More
JULABO™ Refrigerators for Chemicals, KRC Series
Julabo™ Refrigerators for Chemicals is used for storing and cooling of chemicals and hazardous substances. The spark free interior prevents damage caused by spilled or evaporating chemicals.
Brand: JULABO™ 8800718
Description
- With spark free interior
- Storing and cooling of hazardous substances
- Self-protecting control circuit
- Digital temperature display (LED)
- Overload protection for cooling compressor with test button
- Cut-off in case of disturbance with optical alarm signal
- Working and safety sensors are protected against short circuits and disconnection
Specifications
Refrigerator for chemicals | |
-2°C to +12°C | |
PTC | |
4 | |
Designed for storing and cooling hazardous substances | |
KRC180 | |
64 x 60 x 85.5 cm | |
52 cm | |
230 V 50 Hz | |
35 kg |
180 L | |
0.1°C | |
±1°C | |
LED | |
4 Support racks | |
40 x 52 x 70 cm | |
70 cm | |
230 V | |
50 Hz | |
35 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.