missing translation for 'onlineSavingsMsg'
Learn More

Charcoal, Technical, ActIVated for Gas Adsorption, 10-18 Mesh, Granular, Fisher Chemical™

Product Code. 10263610
Click to view available options
Quantity:
500 g
1 kg

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
MDL Number 133992

Specifications

Melting Point 3652°C
Color Black
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 12g/mol
Physical Form Solid
Product Identifier
  • Activated charcoal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.