Learn More
Cesium sulfate, 99.95% (metals basis), Thermo Scientific Chemicals
55.88 € - 212.06 €
Chemical Identifiers
CAS | 10294-54-9 |
---|---|
Molecular Formula | Cs2O4S |
Molecular Weight (g/mol) | 361.87 |
MDL Number | MFCD00010959 |
InChI Key | FLJPGEWQYJVDPF-UHFFFAOYSA-L |
Synonym | cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate |
PubChem CID | 25137 |
SMILES | [Cs+].[Cs+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396717
|
Thermo Scientific Alfa Aesar
010027.14 |
25 g |
55.88 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306727
|
Thermo Scientific Alfa Aesar
010027.22 |
100 g |
212.06 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-54-9 | |
361.87 | |
FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
25137 |
Cs2O4S | |
MFCD00010959 | |
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate | |
[Cs+].[Cs+].[O-]S([O-])(=O)=O |
Specifications
1010°C | |
White | |
99.9% (Metal basis) | |
MFCD00010959 | |
Very soluble in water. Slightly soluble in alcohol,acetone,pyridine. | |
[Cs+].[Cs+].[O-]S([O-])(=O)=O | |
361.87 | |
361.87 | |
White powder | |
Cesium sulfate |
10294-54-9 | |
Powder | |
Cs2O4S | |
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfate | |
FLJPGEWQYJVDPF-UHFFFAOYSA-L | |
dicaesium(1+) sulfate | |
25137 | |
Hygroscopic | |
4.24 g/cm3 at 20°C |
Safety and Handling
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H361f-H373
missing translation for 'einecsNumber' : 233-662-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.