missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium chloride, Reagent Grade, 99.9%, Honeywell™
Reagent Grade, 99.9%
86.37 € - 314.53 €
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
PubChem CID | 24293 |
ChEBI | CHEBI:63039 |
IUPAC Name | cesium;chloride |
SMILES | [Cl-].[Cs+] |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
cesium;chloride |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 | |
[Cl-].[Cs+] |
Specifications
645°C (literature) | |
100.0000% | |
CsCl | |
NONH for all modes of transport | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
cesium;chloride | |
24293 | |
168.36 | |
Reagent | |
3.98 g/cm3 |
7647-17-8 | |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
≥99.9% | |
Poly Bottle | |
Cesium chloride, Reagent Grade, 99.9% |
Safety and Handling
P280
H361
missing translation for 'shelfLife' : 720 days from date of manufacture
missing translation for 'einecsNumber' : 231-600-2
missing translation for 'rtecsNumber' : FK9625000
Recommended Storage : Room Temp
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title