missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride (Crystalline Powder), Fisher BioReagents
For isolation of phage DNA, plasmid DNA, or total RNA
304.00 € - 640.00 €
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride |
PubChem CID | 24293 |
ChEBI | CHEBI:63039 |
IUPAC Name | cesium;chloride |
SMILES | [Cl-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10648783
|
Fisher BioReagents
BP1595-500 |
500 g | Poly Bottle |
304.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386503
|
Fisher BioReagents
BP1595-1 |
1 kg | Poly Bottle |
640.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This economical grade of Cesium Chloride can be used where low levels of metal ion impurities do not interfere. It is recommended for isolating phage DNA, plasmid DNA, or total RNA.Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
cesium;chloride |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
CHEBI:63039 | |
[Cl-].[Cs+] |
Specifications
1290°C | |
7647-17-8 | |
5 to 9 | |
≥99.5 % | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
≥99.5% | |
Cesium Chloride |
646°C | |
White | |
Powder/Solid | |
ClCs | |
15, 2010 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
cesium;chloride | |
24293 | |
168.36 | |
1.364 to 1.374 (at 25°C) | |
DNase-, RNase- and Protease-Free |
Safety and Handling
Recommended Storage : RT
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cesium chloride
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.