Learn More
Certified Reference Flash Point Standard - Cleveland, Lubricant, UKAS ISO 17034 Certified 250mL (nominal value: 256.8 °C), Paragon Scientific™
Flash Point Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Cleveland Open Cup Flash Point ASTM D92
Brand:  Paragon Scientific CRMU-FCLU
Product Code. 16060050
  • 215.00 € / 250mL
Explore more special offers
Description
Description
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified to ISO 17034 under our UKAS accreditation
  • Ensures good interlaboratory correlation
  • Supplied in tamper evident UN compliant security pack
  • Pack size 250mL
  • 12 months shelf life
Specifications
Specifications
250 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.