missing translation for 'onlineSavingsMsg'
Learn More

Certified pH Buffer Solution, UKAS ISO 17025 / ISO 17034 Certified, 20 °C, Paragon Scientific™

Specifically used for verification and calibration checks of pH meter instruments. Color-coded to allow for easy identification of the pH required, and include anti-fungal preservatives / mould inhibitors. pH values are traceable to National Institute Standards and Technology (NIST.)

Brand:  Paragon Scientific PH10-20CF

 View more versions of this product

Product Code. 16030272

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Certified solutions for 20 °C and are accurate within ± 0.01
  • Certified pH buffer solutions are fully traceable to NIST and international protocols
  • Sold as colour-coded solutions for easy-identification purposes
  • Sold with anti-fungal preservatives / mould inhibitors
  • Supplied with certificate, Safety Data Sheet (SDS) and correlation table between pH and temperature
  • Supplied in 500 mL, tamper-evident containers. For other volumes, please contact us direct.
  • 12 month shelf life per solution
Specifications

Specifications

Blue
UKAS ISO 17025 / ISO 17034
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.