Learn More
Cerium oxide, 20% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Widely applied as a solid electrolyte in fuel cells. Also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting.
39.30 € - 353.76 €
Chemical Identifiers
CAS | 1306-38-3 |
---|---|
Molecular Formula | CeO2 |
Molecular Weight (g/mol) | 172.114 |
MDL Number | MFCD00010933 |
InChI Key | CETPSERCERDGAM-UHFFFAOYSA-N |
Synonym | ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline |
PubChem CID | 73963 |
ChEBI | CHEBI:79089 |
IUPAC Name | dioxocerium |
SMILES | O=[Ce]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399257
|
Thermo Scientific Alfa Aesar
040125.22 |
100 g |
39.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309267
|
Thermo Scientific Alfa Aesar
040125.36 |
500 g |
100.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319267
|
Thermo Scientific Alfa Aesar
040125.A3 |
2 kg |
353.76 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-38-3 | |
172.114 | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
73963 | |
dioxocerium |
CeO2 | |
MFCD00010933 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
CHEBI:79089 | |
O=[Ce]=O |
Specifications
1306-38-3 | |
CeO2 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
dioxocerium | |
73963 | |
172.12 | |
Plastic bottle | |
1.22 |
Particles in Liquid | |
MFCD00010933 | |
Immiscible with water. | |
O=[Ce]=O | |
172.114 | |
CHEBI:79089 | |
20% in H2O | |
1.22 | |
Cerium oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-150-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only