missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(IV) sulfate solution 0.1, (0.1N), NIST standard solution ready to use, for volumetric analysis, Fisher Chemical™
117.00 € - 860.00 €
Chemical Identifiers
CAS | 13590-82-4 |
---|---|
Molecular Formula | CeO8S2 |
Molecular Weight (g/mol) | 332.23 |
MDL Number | MFCD00148852 |
InChI Key | VZDYWEUILIUIDF-UHFFFAOYSA-J |
Synonym | ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate |
PubChem CID | 159684 |
IUPAC Name | cerium(4+);disulfate |
SMILES | [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10244340
|
Fisher Chemical
J/3000/15 |
1 L | Amber glass bottle |
117.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10635302
|
Fisher Chemical
J/3000/17 |
2.5 L | Amber glass bottle |
860.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11492704
|
Fisher Chemical
J/3000/24 |
10 L | Bag in box | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
13590-82-4 | |
332.23 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
159684 | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
CeO8S2 | |
MFCD00148852 | |
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
cerium(4+);disulfate |
Specifications
13590-82-4 | |
Liquid | |
MFCD00148852 | |
VZDYWEUILIUIDF-UHFFFAOYSA-J | |
cerium(4+);disulfate | |
159684 | |
Vapor Density: 0.7 |
Yellow | |
CeO8S2 | |
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
332.23 | |
14mmHg at 20°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cerium (IV) sulfate 0.1M in ca. 10% w/v sulfuric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash A§ thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cerium(IV) sulfate solution 0.1, (0.1N), NIST standard solution ready to use, for volumetric analysis, Fisher Chemical™