missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals

25.43 € - 349.74 €

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331099
SDS View Documents
Thermo Scientific Alfa Aesar
011332.09
10 g
25.43 €
10g
Please to purchase this item. Need a web account? Register with us today!
11351099
SDS View Documents
Thermo Scientific Alfa Aesar
011332.22
100 g
109.55 €
100g
Please to purchase this item. Need a web account? Register with us today!
11341099
SDS View Documents
Thermo Scientific Alfa Aesar
011332.36
500 g
349.74 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium(IV) ammonium nitrate is used to offers a novel route to tartronic acid derivatives via facile oxygenation of malonates. It is also used as for oxidative addition reactions of electrophilic radicals to alkenes, enabling intermolecular and intramolecular carbon-carbon and carbon-heteroatom bond formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16774-21-3
H8CeN8O18
MFCD00151121

Specifications

16774-21-3
Crystalline
H8CeN8O18
Soluble in water.
Pungent
Yellow to Orange
99.5% (REO)
MFCD00151121
548.23
Cerium(IV) ammonium nitrate
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c

H272-H290-H302-H314-H317-H335

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 240-827-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cerium(IV) ammonium nitrate, REacton

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Cerium(IV) ammoniumnitrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.