Learn More
Cerium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific™
339.11 €
Chemical Identifiers
CAS | 7758-88-5 |
---|---|
Molecular Formula | CeF3 |
Molecular Weight (g/mol) | 197.111 |
MDL Number | MFCD00010932 |
InChI Key | QCCDYNYSHILRDG-UHFFFAOYSA-K |
Synonym | cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs |
PubChem CID | 24457 |
IUPAC Name | cerium(3+);trifluoride |
SMILES | [F-].[F-].[F-].[Ce+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386918
|
Thermo Scientific Alfa Aesar
013662.30 |
250 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7758-88-5 | |
197.111 | |
QCCDYNYSHILRDG-UHFFFAOYSA-K | |
24457 | |
[F-].[F-].[F-].[Ce+3] |
CeF3 | |
MFCD00010932 | |
cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs | |
cerium(3+);trifluoride |
Specifications
∼2300°C | |
7758-88-5 | |
CeF3 | |
14,1998 | |
Soluble in strong mineral acids. | |
[F-].[F-].[F-].[Ce+3] | |
197.111 | |
197.12 | |
Reacton™ | |
Hygroscopic | |
(REO) |
1460°C | |
Lump | |
MFCD00010932 | |
cerium fluoride, cerous fluoride, cerium 3+ trifluoride, cerium fluorure french, unii-1gct2g09an, cerium +3 cation trifluoride, 1gct2g09an, cerium fluorure, acmc-20alhs | |
QCCDYNYSHILRDG-UHFFFAOYSA-K | |
cerium(3+);trifluoride | |
24457 | |
99.9% | |
Odorless | |
6.16 g/mL | |
Cerium(III) fluoride, Anhydrous |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
missing translation for 'einecsNumber' : 231-841-3
missing translation for 'rtecsNumber' : FK6125000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures