missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) carbonate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

20.90 € - 188.94 €

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
PubChem CID 16212049
IUPAC Name cerium(3+);tricarbonate;hydrate
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10776573
View Documents Promotion Details
Thermo Scientific Acros
379400100
10 g Glass bottle
20.90 €
10g
Please to purchase this item. Need a web account? Register with us today!
10033872
View Documents Promotion Details
Thermo Scientific Acros
379400500
50 g Glass bottle
48.24 €
50g
Please to purchase this item. Need a web account? Register with us today!
10087002
View Documents Promotion Details
Thermo Scientific Acros
379402500
250 g Glass bottle
188.94 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

54451-25-1
460.26
GHLITDDQOMIBFS-UHFFFAOYSA-H
16212049
[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
C3Ce2O9
MFCD00149635
cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
cerium(3+);tricarbonate;hydrate

Specifications

54451-25-1
Crystalline Powder
C3Ce2O9
MFCD00149635
Solubility in water: insoluble
[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
460.26
460.27
Trace Metal Basis
White
99.9%
Ce(CO3)3·xH2O
cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
GHLITDDQOMIBFS-UHFFFAOYSA-H
cerium(3+);tricarbonate;hydrate
16212049
99.9%
Cerium(III) carbonate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only