missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

97.89 € - 1221.08 €

Chemical Identifiers

CAS 14457-87-5
Molecular Formula Br3Ce
Molecular Weight (g/mol) 379.83
MDL Number MFCD00016004
InChI Key MOOUSOJAOQPDEH-UHFFFAOYSA-K
Synonym Cerium tribromide; Cerous bromide
IUPAC Name cerium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Ce+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11323948
SDS View Documents
Thermo Scientific Alfa Aesar
040373.03
1 g
97.89 €
1g
Please to purchase this item. Need a web account? Register with us today!
11333948
SDS View Documents
Thermo Scientific Alfa Aesar
040373.06
5 g
295.47 €
5g
Please to purchase this item. Need a web account? Register with us today!
11420271
SDS View Documents
Thermo Scientific Alfa Aesar
040373.14
25 g
1221.08 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium(III) bromide is used as an additive for chemical reagent. Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 238-447-0

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.