Learn More
OHAUS™ Centrifuge Rack
Adapter accessory for Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Brand:  OHAUS™ 30553128
 View more versions of this product
Product Code. 16602774
  • 294.00 € / Pack of 2
Explore more special offers
Description
Description
  • Adapter for centrifuges:
    • Centrifuge Multi FC5916R
    • Centrifuge Multi FC5916
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 x 50 mL
Centrifuge Multi FC5916R and Centrifuge Multi FC5916
Depth 29 mm
FA x2